Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR27
Entrez Gene ID
253769
Gene Name
WD repeat domain 27
Gene Aliases
-
Chromosome Location
Chr.6: 169426420 - 169702071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202550.1 NP_001189479.1 20 - 21 2567 113
NM_182552.4 NP_872358.4 23 - 24 2948 113
XM_011535682.2 XP_011533984.1 24 - 25 3138 113
XM_011535684.2 XP_011533986.1 24 - 25 3138 113
XM_011535685.2 XP_011533987.1 24 - 25 3138 113
XM_011535687.2 XP_011533989.1 24 - 25 3138 113
XM_011535688.2 XP_011533990.1 24 - 25 3138 113
XM_011535689.2 XP_011533991.1 24 - 25 3138 113
XM_011535690.2 XP_011533992.1 24 - 25 3138 113
XM_011535691.2 XP_011533993.1 24 - 25 3138 113
XM_011535692.2 XP_011533994.1 24 - 25 3138 113
XM_011535694.2 XP_011533996.1 23 - 24 3040 113
XM_017010658.1 XP_016866147.1 24 - 25 3138 113
XM_017010659.1 XP_016866148.1 24 - 25 3138 113
XM_017010660.1 XP_016866149.1 24 - 25 3138 113
XM_017010661.1 XP_016866150.1 24 - 25 3138 113
XM_017010662.1 XP_016866151.1 24 - 25 3138 113
XM_017010669.1 XP_016866158.1 18 - 19 2383 113
XM_017010670.1 XP_016866159.1 23 - 24 3004 113
XM_017010671.1 XP_016866160.1 22 - 23 2906 113
XM_017010673.1 XP_016866162.1 21 - 22 2784 113
GenBank mRNA AK131435.1 - 12 - 13 3097 113
BC131633.1 - 21 - 22 2090 113
BX648854.1 - 18 - 19 1809 113

View Full Product Details