The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321471.1 | NP_001308400.1 | 4 - 5 | 498 | 98 |
NM_001321473.1 | NP_001308402.1 | 3 - 4 | 366 | 98 | |
NM_005119.3 | NP_005110.2 | 3 - 4 | 362 | 98 | |
XM_005271371.4 | XP_005271428.1 | 1 - 2 | 142 | 98 | |
GenBank mRNA | AF117756.1 | - | 3 - 4 | 340 | 98 |
BC002501.2 | - | 3 - 4 | 333 | 98 | |
BC010997.2 | - | 3 - 4 | 335 | 98 | |
BC019076.1 | - | 3 - 4 | 328 | 98 | |
BC037554.2 | - | 2 - 3 | 274 | 98 | |
BC054046.1 | - | 3 - 4 | 389 | 98 | |
BC065519.1 | - | 3 - 4 | 338 | 98 | |
BC112330.1 | - | 3 - 4 | 324 | 98 | |
BC112350.1 | - | 4 - 5 | 456 | 98 | |
BG572651.1 | - | 3 - 4 | 363 | 98 | |
HY001123.1 | - | 3 - 4 | 366 | 98 |