Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYNC2H1
Entrez Gene ID
79659
Gene Name
dynein cytoplasmic 2 heavy chain 1
Gene Aliases
ATD3, DHC1b, DHC2, DNCH2, DYH1B, SRPS2B, SRTD3, hdhc11
Chromosome Location
Chr.11: 103109431 - 103479863 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080463.1 NP_001073932.1 45 - 46 7587 105
NM_001377.2 NP_001368.2 45 - 46 7587 105
XM_006718903.2 XP_006718966.1 45 - 46 7587 105
XM_017018291.1 XP_016873780.1 45 - 46 7587 105
XM_017018292.1 XP_016873781.1 42 - 43 6841 105
GenBank mRNA AB290167.1 - 45 - 46 7641 105

View Full Product Details