Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYNC2H1
Entrez Gene ID
79659
Gene Name
dynein cytoplasmic 2 heavy chain 1
Gene Aliases
ATD3, DHC1b, DHC2, DNCH2, DYH1B, SRPS2B, SRTD3, hdhc11
Chromosome Location
Chr.11: 103109431 - 103479863 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080463.1 NP_001073932.1 60 - 61 9585 144
NM_001377.2 NP_001368.2 60 - 61 9585 144
XM_006718903.2 XP_006718966.1 60 - 61 9564 144
XM_017018291.1 XP_016873780.1 60 - 61 9585 144
XM_017018292.1 XP_016873781.1 57 - 58 8839 144
GenBank mRNA AB082528.2 - 12 - 13 1576 144
AB290167.1 - 60 - 61 9639 144
AK131453.1 - 5 - 6 579 144
BX538093.1 - 13 - 14 1695 144
BX649161.1 - 8 - 9 960 144

View Full Product Details