Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO8
Entrez Gene ID
26269
Gene Name
F-box protein 8
Gene Aliases
DC10, FBS, FBX8
Chromosome Location
Chr.4: 174236658 - 174284251 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012180.2 NP_036312.2 4 - 5 1425 149
XM_017008020.1 XP_016863509.1 4 - 5 689 149
GenBank mRNA AF115514.1 - 4 - 5 809 149
AF174596.1 - 4 - 5 641 149
AF201932.1 - 4 - 5 834 149
AF233224.1 - 4 - 5 680 149
AK026319.1 - 4 - 5 848 149
AK123740.1 - 4 - 5 1438 149
AY358721.1 - 4 - 5 811 149
BC014679.2 - 4 - 5 827 149
BC014903.1 - 4 - 5 1425 149
BC040456.1 - 5 - 6 718 149
KU178588.1 - 3 - 4 575 149

View Full Product Details