The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006576.3 | NP_006567.3 | 16 - 17 | 2181 | 92 |
XM_017018710.1 | XP_016874199.1 | 17 - 18 | 2247 | 92 | |
XM_017018711.1 | XP_016874200.1 | 17 - 18 | 4993 | 92 | |
XM_017018712.1 | XP_016874201.1 | 17 - 18 | 4936 | 92 | |
XM_017018713.1 | XP_016874202.1 | 17 - 18 | 4596 | 92 | |
XM_017018714.1 | XP_016874203.1 | 9 - 10 | 1396 | 92 | |
XM_017018715.1 | XP_016874204.1 | 9 - 10 | 1270 | 92 | |
GenBank mRNA | AF041449.1 | - | 16 - 17 | 2181 | 92 |
AK314362.1 | - | 16 - 17 | 2583 | 92 | |
BC111730.1 | - | 17 - 18 | 2285 | 92 |