Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NXF3
Entrez Gene ID
56000
Gene Name
nuclear RNA export factor 3
Gene Aliases
-
Chromosome Location
Chr.X: 103075821 - 103093094 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022052.1 NP_071335.1 8 - 9 854 70
GenBank mRNA AF346619.1 - 8 - 9 854 70
AJ277527.1 - 8 - 9 854 70
AJ277660.1 - 5 - 6 412 70
AK302447.1 - 8 - 9 855 70
AK302586.1 - 7 - 8 820 70
BC031616.1 - 8 - 9 847 70

View Full Product Details