Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOM1
Entrez Gene ID
64434
Gene Name
nucleolar protein with MIF4G domain 1
Gene Aliases
C7orf3, PPP1R113, SGD1
Chromosome Location
Chr.7: 156949723 - 156973182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138400.1 NP_612409.1 7 - 8 2049 152
GenBank mRNA AY576779.1 - 7 - 8 2049 152
BC007902.2 - 3 - 4 340 152

View Full Product Details