The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004584.2 | NP_004575.1 | 3 - 4 | 322 | 98 |
XM_006718652.3 | XP_006718715.1 | 1 - 2 | 534 | 98 | |
XM_017018097.1 | XP_016873586.1 | 3 - 4 | 298 | 98 | |
GenBank mRNA | AK315348.1 | - | 3 - 4 | 261 | 98 |
BC014848.1 | - | 3 - 4 | 322 | 98 | |
CR536508.1 | - | 3 - 4 | 229 | 98 | |
U53174.1 | - | 3 - 4 | 305 | 98 |