The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304815.1 | NP_001291744.1 | 13 - 14 | 5804 | 60 |
NM_015125.4 | NP_055940.3 | 12 - 13 | 3049 | 60 | |
XM_005258673.2 | XP_005258730.1 | 12 - 13 | 3141 | 60 | |
XM_005258674.2 | XP_005258731.1 | 12 - 13 | 3141 | 60 | |
XM_005258675.2 | XP_005258732.1 | 12 - 13 | 3141 | 60 | |
XM_011526660.2 | XP_011524962.1 | 13 - 14 | 5846 | 60 | |
XM_011526661.2 | XP_011524963.1 | 13 - 14 | 5846 | 60 | |
XM_011526662.2 | XP_011524964.1 | 13 - 14 | 5846 | 60 | |
XM_011526663.2 | XP_011524965.1 | 13 - 14 | 5846 | 60 | |
XM_011526664.2 | XP_011524966.1 | 13 - 14 | 5846 | 60 | |
XM_011526665.2 | XP_011524967.1 | 13 - 14 | 5846 | 60 | |
XM_011526666.2 | XP_011524968.1 | 13 - 14 | 5846 | 60 | |
GenBank mRNA | AB002304.1 | - | 9 - 10 | 2540 | 60 |
AF363689.1 | - | 12 - 13 | 3049 | 60 |