The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001221.3 | NP_001212.2 | 12 - 13 | 1811 | 77 |
NM_172115.2 | NP_742113.1 | 12 - 13 | 1811 | 77 | |
NM_172127.2 | NP_742125.1 | 12 - 13 | 1811 | 77 | |
NM_172128.2 | NP_742126.1 | 12 - 13 | 1811 | 77 | |
XM_006714330.2 | XP_006714393.1 | 12 - 13 | 1650 | 77 | |
GenBank mRNA | AF071569.1 | - | 12 - 13 | 952 | 77 |
AK025368.1 | - | 1 - 2 | 62 | 77 | |
AK055642.1 | - | 12 - 13 | 1329 | 77 | |
BC032784.1 | - | 12 - 13 | 1014 | 77 |