Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAMK2D
Entrez Gene ID
817
Gene Name
calcium/calmodulin dependent protein kinase II delta
Gene Aliases
CAMKD
Chromosome Location
Chr.4: 113451032 - 113761927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001221.3 NP_001212.2 17 - 18 2293 113
NM_001321566.1 NP_001308495.1 18 - 19 2335 113
NM_001321569.1 NP_001308498.1 19 - 20 2395 113
NM_001321571.1 NP_001308500.1 19 - 20 2395 113
NM_001321573.1 NP_001308502.1 19 - 20 2368 113
NM_001321574.1 NP_001308503.1 17 - 18 2290 113
NM_001321580.1 NP_001308509.1 18 - 19 2353 113
NM_001321588.1 NP_001308517.1 18 - 19 2332 113
XM_005263253.3 XP_005263310.1 19 - 20 2210 113
XM_005263254.3 XP_005263311.1 19 - 20 2231 113
XM_011532289.1 XP_011530591.1 18 - 19 2144 113
XM_011532290.1 XP_011530592.1 18 - 19 2141 113
XM_011532292.2 XP_011530594.1 15 - 16 1245 113
XM_011532295.1 XP_011530597.1 18 - 19 2165 113
XM_017008672.1 XP_016864161.1 18 - 19 2174 113
XM_017008673.1 XP_016864162.1 18 - 19 2171 113
XM_017008675.1 XP_016864164.1 20 - 21 2335 113
XM_017008676.1 XP_016864165.1 19 - 20 1571 113
GenBank mRNA AF071569.1 - 17 - 18 1434 113
AK294517.1 - 19 - 20 1686 113
AL120285.1 - 2 - 3 246 113

View Full Product Details