The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012262.3 | NP_036394.1 | 6 - 7 | 1234 | 90 |
GenBank mRNA | AB007917.1 | - | 6 - 7 | 1131 | 90 |
AB024568.1 | - | 6 - 7 | 1203 | 90 | |
AK023260.1 | - | 6 - 7 | 1207 | 90 |