The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258452.1 | NP_001245381.1 | 20 - 21 | 3044 | 84 |
NM_001258453.1 | NP_001245382.1 | 20 - 21 | 3065 | 84 | |
NM_032807.4 | NP_116196.3 | 20 - 21 | 3081 | 84 | |
NM_178150.2 | NP_835363.1 | 19 - 20 | 2967 | 84 | |
XM_011519748.2 | XP_011518050.1 | 21 - 22 | 3661 | 84 | |
XM_017016842.1 | XP_016872331.1 | 21 - 22 | 4259 | 84 | |
XM_017016843.1 | XP_016872332.1 | 22 - 23 | 3643 | 84 | |
XM_017016844.1 | XP_016872333.1 | 20 - 21 | 3028 | 84 | |
XM_017016845.1 | XP_016872334.1 | 20 - 21 | 3385 | 84 | |
XM_017016846.1 | XP_016872335.1 | 18 - 19 | 2786 | 84 | |
GenBank mRNA | AF380349.1 | - | 20 - 21 | 3081 | 84 |
AF454502.1 | - | 19 - 20 | 2908 | 84 | |
AF456237.1 | - | 19 - 20 | 2923 | 84 | |
AK027381.1 | - | 10 - 11 | 1087 | 84 | |
AK027496.1 | - | 16 - 17 | 2050 | 84 | |
AK122753.1 | - | 20 - 21 | 3044 | 84 | |
AK226128.1 | - | 19 - 20 | 3491 | 84 | |
AK315173.1 | - | 18 - 19 | 2683 | 84 | |
AL133069.1 | - | 5 - 6 | 540 | 84 | |
BC012762.2 | - | 6 - 7 | 709 | 84 | |
BC020266.1 | - | 17 - 18 | 2475 | 84 | |
BC032674.1 | - | 17 - 18 | 2419 | 84 | |
BC080629.1 | - | 17 - 18 | 2630 | 84 | |
BC110884.1 | - | 20 - 21 | 2990 | 84 | |
BC113375.1 | - | 19 - 20 | 2914 | 84 | |
BC113377.1 | - | 19 - 20 | 2914 | 84 |