The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258452.1 | NP_001245381.1 | 4 - 5 | 1050 | 72 |
NM_001258453.1 | NP_001245382.1 | 5 - 6 | 1122 | 72 | |
NM_032807.4 | NP_116196.3 | 5 - 6 | 1138 | 72 | |
NM_178150.2 | NP_835363.1 | 4 - 5 | 1024 | 72 | |
XM_011519748.2 | XP_011518050.1 | 6 - 7 | 1718 | 72 | |
XM_017016842.1 | XP_016872331.1 | 5 - 6 | 2265 | 72 | |
XM_017016843.1 | XP_016872332.1 | 6 - 7 | 1649 | 72 | |
XM_017016844.1 | XP_016872333.1 | 4 - 5 | 1034 | 72 | |
XM_017016845.1 | XP_016872334.1 | 4 - 5 | 1391 | 72 | |
XM_017016846.1 | XP_016872335.1 | 3 - 4 | 880 | 72 | |
GenBank mRNA | AF380349.1 | - | 5 - 6 | 1138 | 72 |
AF454502.1 | - | 4 - 5 | 968 | 72 | |
AF456237.1 | - | 4 - 5 | 980 | 72 | |
AK027496.1 | - | 1 - 2 | 110 | 72 | |
AK095343.1 | - | 4 - 5 | 998 | 72 | |
AK122753.1 | - | 4 - 5 | 1050 | 72 | |
AK226128.1 | - | 4 - 5 | 1548 | 72 | |
AK315173.1 | - | 3 - 4 | 740 | 72 | |
BC006430.1 | - | 2 - 3 | 715 | 72 | |
BC020266.1 | - | 2 - 3 | 532 | 72 | |
BC032674.1 | - | 2 - 3 | 476 | 72 | |
BC080629.1 | - | 2 - 3 | 687 | 72 | |
BC110884.1 | - | 5 - 6 | 1047 | 72 | |
BC113375.1 | - | 4 - 5 | 971 | 72 | |
BC113377.1 | - | 4 - 5 | 971 | 72 |