The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005521.3 | NP_005512.1 | 2 - 3 | 1012 | 54 |
XM_011539744.2 | XP_011538046.1 | 2 - 3 | 1564 | 54 | |
GenBank mRNA | M62626.1 | - | 2 - 3 | 1792 | 54 |
M75952.1 | - | 2 - 3 | 786 | 54 | |
S38742.1 | - | 2 - 3 | 872 | 54 |