Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKRD2
Entrez Gene ID
26287
Gene Name
ankyrin repeat domain 2
Gene Aliases
ARPP
Chromosome Location
Chr.10: 97572441 - 97583884 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129981.2 NP_001123453.1 3 - 4 696 80
NM_001291218.1 NP_001278147.1 3 - 4 696 80
NM_001291219.1 NP_001278148.1 3 - 4 696 80
NM_020349.3 NP_065082.2 3 - 4 696 80
GenBank mRNA AB058599.1 - 3 - 4 358 80
AJ304805.1 - 3 - 4 358 80
AJ583444.1 - 3 - 4 696 80
AK056990.1 - 3 - 4 638 80
AK314763.1 - 3 - 4 358 80
BC020817.1 - 3 - 4 358 80
BC107759.1 - 3 - 4 599 80

View Full Product Details