The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000090.3 | NP_000081.1 | 2 - 3 | 402 | 79 |
GenBank mRNA | AK308010.1 | - | 2 - 3 | 402 | 79 |
BC028178.1 | - | 2 - 3 | 400 | 79 | |
BP374999.1 | - | 2 - 3 | 402 | 79 | |
X07240.1 | - | 2 - 3 | 285 | 79 | |
X14420.1 | - | 2 - 3 | 387 | 79 |