Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL3A1
Entrez Gene ID
1281
Gene Name
collagen type III alpha 1 chain
Gene Aliases
EDS4A
Chromosome Location
Chr.2: 188974373 - 189012746 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000090.3 NP_000081.1 8 - 9 807 94
GenBank mRNA BC028178.1 - 8 - 9 805 94
X14420.1 - 8 - 9 792 94
X15332.1 - 4 - 5 249 94

View Full Product Details