The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080400.1 | NP_001073869.1 | 4 - 5 | 3655 | 69 |
XM_006722866.2 | XP_006722929.1 | 6 - 7 | 5616 | 69 | |
XM_006722868.3 | XP_006722931.1 | 6 - 7 | 5612 | 69 | |
XM_011528233.2 | XP_011526535.1 | 7 - 8 | 5798 | 69 | |
XM_017027192.1 | XP_016882681.1 | 7 - 8 | 5801 | 69 | |
XM_017027193.1 | XP_016882682.1 | 7 - 8 | 5748 | 69 | |
XM_017027194.1 | XP_016882683.1 | 7 - 8 | 5783 | 69 | |
GenBank mRNA | AB067468.1 | - | - | 3629 | 69 |
AK295236.1 | - | 2 - 3 | 580 | 69 | |
BC153818.1 | - | 3 - 4 | 1511 | 69 |