The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016329.3 | NP_057413.2 | 6 - 7 | 1088 | 77 |
XM_005265221.3 | XP_005265278.1 | 6 - 7 | 1204 | 77 | |
XM_006713203.3 | XP_006713266.1 | 7 - 8 | 1326 | 77 | |
XM_006713204.3 | XP_006713267.1 | 6 - 7 | 1026 | 77 | |
XM_011533824.2 | XP_011532126.1 | 7 - 8 | 1202 | 77 | |
XM_011533825.2 | XP_011532127.1 | 7 - 8 | 1049 | 77 | |
GenBank mRNA | AB189472.1 | - | 5 - 6 | 712 | 77 |
AF168132.1 | - | 6 - 7 | 1025 | 77 | |
AK313965.1 | - | 6 - 7 | 1083 | 77 | |
AL080140.1 | - | 3 - 4 | 366 | 77 | |
BC014614.2 | - | 6 - 7 | 972 | 77 |