The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016329.3 | NP_057413.2 | 8 - 9 | 1278 | 84 |
XM_005265221.3 | XP_005265278.1 | 8 - 9 | 1394 | 84 | |
XM_006713203.3 | XP_006713266.1 | 9 - 10 | 1516 | 84 | |
XM_006713204.3 | XP_006713267.1 | 8 - 9 | 1216 | 84 | |
XM_011533824.2 | XP_011532126.1 | 9 - 10 | 1392 | 84 | |
XM_011533825.2 | XP_011532127.1 | 9 - 10 | 1239 | 84 | |
GenBank mRNA | AB189472.1 | - | 7 - 8 | 902 | 84 |
AF168132.1 | - | 8 - 9 | 1215 | 84 | |
AK313965.1 | - | 8 - 9 | 1273 | 84 | |
AL080140.1 | - | 5 - 6 | 556 | 84 | |
AL523328.3 | - | 3 - 4 | 305 | 84 | |
BC014614.2 | - | 8 - 9 | 1162 | 84 |