Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASPH
Entrez Gene ID
444
Gene Name
aspartate beta-hydroxylase
Gene Aliases
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
Chromosome Location
Chr.8: 61500556 - 61714640 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164750.1 NP_001158222.1 24 - 25 2202 63
NM_004318.3 NP_004309.2 24 - 25 2397 63
XM_005251235.2 XP_005251292.1 25 - 26 2411 63
XM_005251236.2 XP_005251293.1 25 - 26 2408 63
XM_005251238.2 XP_005251295.1 24 - 25 2366 63
XM_005251239.2 XP_005251296.1 24 - 25 2354 63
XM_005251240.1 XP_005251297.1 25 - 26 2189 63
XM_005251242.2 XP_005251299.1 23 - 24 2309 63
XM_005251243.2 XP_005251300.1 24 - 25 2282 63
XM_005251244.1 XP_005251301.1 24 - 25 2129 63
XM_005251246.2 XP_005251303.1 23 - 24 2237 63
XM_005251247.2 XP_005251304.1 23 - 24 2225 63
XM_005251248.1 XP_005251305.1 23 - 24 1995 63
XM_005251250.2 XP_005251307.1 22 - 23 2180 63
XM_017013429.1 XP_016868918.1 24 - 25 2363 63
XM_017013434.1 XP_016868923.1 24 - 25 2351 63
XM_017013436.1 XP_016868925.1 23 - 24 2324 63
XM_017013438.1 XP_016868927.1 24 - 25 2279 63
XM_017013439.1 XP_016868928.1 24 - 25 2049 63
XM_017013441.1 XP_016868930.1 23 - 24 2234 63
XM_017013442.1 XP_016868931.1 23 - 24 2222 63
XM_017013443.1 XP_016868932.1 22 - 23 2177 63
XM_017013444.1 XP_016868933.1 23 - 24 1923 63
GenBank mRNA AK304314.1 - 24 - 25 2158 63
U03109.1 - 24 - 25 2202 63

View Full Product Details