The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305005.2 | NP_001291934.1 | 7 - 8 | 1191 | 69 |
NM_001305006.2 | NP_001291935.1 | 6 - 7 | 965 | 69 | |
NM_001305007.2 | NP_001291936.1 | 7 - 8 | 1157 | 69 | |
NM_021815.4 | NP_068587.1 | 7 - 8 | 1194 | 69 | |
XM_011511580.2 | XP_011509882.1 | 5 - 6 | 745 | 69 | |
XM_017004628.1 | XP_016860117.1 | 5 - 6 | 1965 | 69 | |
XM_017004629.1 | XP_016860118.1 | 5 - 6 | 4473 | 69 | |
GenBank mRNA | AB043997.1 | - | 7 - 8 | 1169 | 69 |
AF276871.1 | - | 6 - 7 | 893 | 69 | |
AJ401466.1 | - | 6 - 7 | 911 | 69 | |
AK300801.1 | - | 6 - 7 | 955 | 69 | |
AK315280.1 | - | 7 - 8 | 1183 | 69 | |
BC111524.1 | - | 7 - 8 | 1099 | 69 | |
BC111525.1 | - | 7 - 8 | 1141 | 69 |