Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC5A7
Entrez Gene ID
60482
Gene Name
solute carrier family 5 member 7
Gene Aliases
CHT, CHT1, HMN7A
Chromosome Location
Chr.2: 107986514 - 108014497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305005.2 NP_001291934.1 8 - 9 1411 67
NM_001305006.2 NP_001291935.1 7 - 8 1185 67
NM_001305007.2 NP_001291936.1 8 - 9 1377 67
NM_021815.4 NP_068587.1 8 - 9 1414 67
XM_011511580.2 XP_011509882.1 6 - 7 965 67
XM_017004628.1 XP_016860117.1 6 - 7 2185 67
XM_017004629.1 XP_016860118.1 6 - 7 4693 67
GenBank mRNA AB043997.1 - 8 - 9 1389 67
AF276871.1 - 7 - 8 1113 67
AJ401466.1 - 7 - 8 1131 67
AK300801.1 - 7 - 8 1175 67
AK315280.1 - 8 - 9 1403 67
BC111524.1 - 8 - 9 1319 67
BC111525.1 - 8 - 9 1361 67

View Full Product Details