The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291824.1 | NP_001278753.1 | 7 - 8 | 1087 | 89 |
NM_001291825.1 | NP_001278754.1 | 6 - 7 | 1100 | 89 | |
NM_001291826.1 | NP_001278755.1 | 6 - 7 | 995 | 89 | |
NM_001291828.1 | NP_001278757.1 | 4 - 5 | 697 | 89 | |
NM_004810.3 | NP_004801.1 | 7 - 8 | 1073 | 89 | |
XM_006724376.3 | XP_006724439.1 | 6 - 7 | 990 | 89 | |
GenBank mRNA | AF042380.1 | - | 7 - 8 | 1002 | 89 |
AF090456.1 | - | 7 - 8 | 964 | 89 | |
AF121002.1 | - | 6 - 7 | 1107 | 89 | |
AF129476.1 | - | 7 - 8 | 992 | 89 | |
AF236119.1 | - | 7 - 8 | 1033 | 89 | |
AF236120.1 | - | 6 - 7 | 810 | 89 | |
AJ011736.1 | - | 7 - 8 | 1018 | 89 | |
AK303280.1 | - | 6 - 7 | 980 | 89 | |
AK303332.1 | - | 5 - 6 | 887 | 89 | |
AK303470.1 | - | 4 - 5 | 688 | 89 | |
AK303512.1 | - | 4 - 5 | 742 | 89 | |
AK303657.1 | - | 6 - 7 | 952 | 89 | |
AK312830.1 | - | 7 - 8 | 1072 | 89 | |
AY069959.1 | - | 7 - 8 | 1073 | 89 | |
BC025692.1 | - | 7 - 8 | 1007 | 89 | |
BC026002.1 | - | 6 - 7 | 1069 | 89 | |
CR456498.1 | - | 7 - 8 | 871 | 89 | |
CR536524.1 | - | 6 - 7 | 810 | 89 | |
CR541849.1 | - | 6 - 7 | 810 | 89 | |
Y18051.1 | - | 7 - 8 | 846 | 89 |