The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020397.3 | NP_065130.1 | 7 - 8 | 1029 | 93 |
NM_153498.3 | NP_705718.1 | 7 - 8 | 1029 | 93 | |
XM_006717481.3 | XP_006717544.1 | 7 - 8 | 1293 | 93 | |
XM_006717482.3 | XP_006717545.1 | 7 - 8 | 1058 | 93 | |
XM_006717483.3 | XP_006717546.1 | 7 - 8 | 1020 | 93 | |
XM_011519591.2 | XP_011517893.1 | 7 - 8 | 1466 | 93 | |
XM_011519592.2 | XP_011517894.1 | 6 - 7 | 1658 | 93 | |
XM_011519593.2 | XP_011517895.1 | 6 - 7 | 1706 | 93 | |
XM_011519594.2 | XP_011517896.1 | 5 - 6 | 647 | 93 | |
XM_011519595.2 | XP_011517897.1 | 6 - 7 | 1520 | 93 | |
XM_017016438.1 | XP_016871927.1 | 8 - 9 | 1324 | 93 | |
XM_017016439.1 | XP_016871928.1 | 9 - 10 | 1155 | 93 | |
GenBank mRNA | AB081726.1 | - | 7 - 8 | 985 | 93 |
AF286366.1 | - | 7 - 8 | 840 | 93 | |
AK289393.1 | - | 8 - 9 | 840 | 93 | |
BC035745.1 | - | 7 - 8 | 846 | 93 |