Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SLC4A9
- Entrez Gene ID
-
83697
- Gene Name
- solute carrier family 4 member 9
- Gene Aliases
-
AE4
- Chromosome Location
-
Chr.5: 140359645 - 140375177 on Build GRCh38
View Full Product Details