The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012302.2 | NP_001012302.2 | 17 - 18 | 1576 | 65 |
XM_011520053.2 | XP_011518355.1 | 17 - 18 | 1672 | 65 | |
XM_017017644.1 | XP_016873133.1 | 18 - 19 | 1706 | 65 | |
XM_017017645.1 | XP_016873134.1 | 18 - 19 | 1658 | 65 | |
XM_017017646.1 | XP_016873135.1 | 17 - 18 | 1595 | 65 | |
XM_017017647.1 | XP_016873136.1 | 17 - 18 | 1547 | 65 | |
GenBank mRNA | AK026923.1 | - | 17 - 18 | 1523 | 65 |
AK074088.1 | - | 12 - 13 | 3358 | 65 | |
AK096874.1 | - | 8 - 9 | 864 | 65 | |
AL832289.1 | - | 7 - 8 | 765 | 65 | |
BC128557.1 | - | 17 - 18 | 1544 | 65 | |
BC144415.1 | - | 16 - 17 | 1602 | 65 |