Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HUWE1
Entrez Gene ID
10075
Gene Name
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
Gene Aliases
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1
Chromosome Location
Chr.X: 53532096 - 53686729 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031407.6 NP_113584.3 75 - 76 12039 75
XM_005261965.3 XP_005262022.1 74 - 75 12106 75
XM_011530747.1 XP_011529049.1 76 - 77 12282 75
XM_011530751.2 XP_011529053.1 75 - 76 12150 75
XM_017029191.1 XP_016884680.1 74 - 75 15034 75
XM_017029192.1 XP_016884681.1 74 - 75 15031 75
XM_017029193.1 XP_016884682.1 74 - 75 15013 75
XM_017029194.1 XP_016884683.1 74 - 75 14989 75
XM_017029195.1 XP_016884684.1 74 - 75 14986 75
XM_017029196.1 XP_016884685.1 74 - 75 14983 75
XM_017029197.1 XP_016884686.1 73 - 74 14935 75
XM_017029198.1 XP_016884687.1 75 - 76 12376 75
XM_017029199.1 XP_016884688.1 76 - 77 12303 75
XM_017029200.1 XP_016884689.1 75 - 76 12173 75
XM_017029201.1 XP_016884690.1 75 - 76 12190 75
XM_017029202.1 XP_016884691.1 75 - 76 12166 75
XM_017029203.1 XP_016884692.1 75 - 76 12172 75
XM_017029204.1 XP_016884693.1 73 - 74 14784 75
XM_017029205.1 XP_016884694.1 72 - 73 14662 75
XM_017029206.1 XP_016884695.1 71 - 72 14632 75
GenBank mRNA AB002310.3 - 44 - 45 8090 75
AB071605.1 - 48 - 49 8739 75
AF161390.1 - 21 - 22 4286 75
AY772009.1 - 73 - 74 11684 75
AY929612.1 - 72 - 73 11636 75
BC072421.1 - 4 - 5 448 75
DQ097177.1 - 75 - 76 12038 75

View Full Product Details