Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HUWE1
Entrez Gene ID
10075
Gene Name
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
Gene Aliases
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1
Chromosome Location
Chr.X: 53532096 - 53686729 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031407.6 NP_113584.3 12 - 13 1267 117
XM_005261965.3 XP_005262022.1 11 - 12 1334 117
XM_011530747.1 XP_011529049.1 12 - 13 1261 117
XM_011530751.2 XP_011529053.1 11 - 12 1129 117
XM_017029193.1 XP_016884682.1 10 - 11 3992 117
XM_017029205.1 XP_016884694.1 10 - 11 3992 117
GenBank mRNA AY772009.1 - 10 - 11 912 117
AY929612.1 - 9 - 10 864 117
DQ097177.1 - 12 - 13 1266 117

View Full Product Details