Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HUWE1
Entrez Gene ID
10075
Gene Name
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
Gene Aliases
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1
Chromosome Location
Chr.X: 53532096 - 53686729 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031407.6 NP_113584.3 9 - 10 1043 136
XM_005261965.3 XP_005262022.1 8 - 9 1110 136
XM_011530747.1 XP_011529049.1 9 - 10 1037 136
XM_011530751.2 XP_011529053.1 8 - 9 905 136
XM_017029191.1 XP_016884680.1 7 - 8 3768 136
XM_017029192.1 XP_016884681.1 7 - 8 3768 136
XM_017029193.1 XP_016884682.1 7 - 8 3768 136
XM_017029194.1 XP_016884683.1 7 - 8 3768 136
XM_017029195.1 XP_016884684.1 7 - 8 3768 136
XM_017029196.1 XP_016884685.1 7 - 8 3768 136
XM_017029197.1 XP_016884686.1 7 - 8 3768 136
XM_017029198.1 XP_016884687.1 8 - 9 1110 136
XM_017029199.1 XP_016884688.1 9 - 10 1037 136
XM_017029200.1 XP_016884689.1 8 - 9 907 136
XM_017029201.1 XP_016884690.1 8 - 9 924 136
XM_017029202.1 XP_016884691.1 8 - 9 900 136
XM_017029203.1 XP_016884692.1 8 - 9 906 136
XM_017029204.1 XP_016884693.1 7 - 8 3767 136
XM_017029205.1 XP_016884694.1 7 - 8 3768 136
XM_017029206.1 XP_016884695.1 7 - 8 3768 136
GenBank mRNA AY772009.1 - 7 - 8 688 136
AY929612.1 - 6 - 7 640 136
DQ097177.1 - 9 - 10 1042 136

View Full Product Details