The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018085.4 | NP_060555.2 | 11 - 12 | 1269 | 157 |
GenBank mRNA | AF410465.1 | - | 11 - 12 | 1219 | 157 |
AK027532.1 | - | 2 - 3 | 133 | 157 | |
AK074740.1 | - | 2 - 3 | 150 | 157 |