Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IPO9
Entrez Gene ID
55705
Gene Name
importin 9
Gene Aliases
Imp9
Chromosome Location
Chr.1: 201829160 - 201884294 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA DB232332.1 - 2 - 3 329 86

View Full Product Details