The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317929.1 | NP_001304858.1 | 4 - 5 | 1363 | 66 |
NM_024330.1 | NP_077306.1 | 4 - 5 | 1363 | 66 | |
GenBank mRNA | AJ577572.1 | - | 4 - 5 | 1656 | 66 |
AK027499.1 | - | 4 - 5 | 1267 | 66 | |
AK075377.1 | - | 4 - 5 | 1356 | 66 | |
AK123997.1 | - | 5 - 6 | 827 | 66 | |
AK125102.1 | - | 4 - 5 | 2301 | 66 | |
AY358409.1 | - | 4 - 5 | 1351 | 66 | |
BC003041.1 | - | 4 - 5 | 1363 | 66 | |
BC003654.2 | - | 3 - 4 | 412 | 66 | |
BC009916.2 | - | 4 - 5 | 1363 | 66 | |
BC029792.1 | - | 4 - 5 | 1231 | 66 |