Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC27A3
Entrez Gene ID
11000
Gene Name
solute carrier family 27 member 3
Gene Aliases
ACSVL3, FATP3, VLCS-3
Chromosome Location
Chr.1: 153774989 - 153780157 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024330.1 NP_077306.1 9 - 10 2078 65
GenBank mRNA AJ577572.1 - 9 - 10 2371 65
AK027499.1 - 9 - 10 1982 65
AK075377.1 - 9 - 10 2071 65
AK125102.1 - 9 - 10 3015 65
AY358409.1 - 9 - 10 2066 65
BC003041.1 - 9 - 10 2078 65
BC009916.2 - 9 - 10 2078 65
BC029792.1 - 9 - 10 1946 65

View Full Product Details