The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020728.2 | NP_065779.1 | 15 - 16 | 1790 | 151 |
GenBank mRNA | AB033054.2 | - | 16 - 17 | 1576 | 151 |
AK001181.1 | - | 4 - 5 | 279 | 151 | |
AK126214.1 | - | 15 - 16 | 1435 | 151 | |
AK308824.1 | - | 15 - 16 | 1398 | 151 | |
AL833233.1 | - | 15 - 16 | 1399 | 151 | |
AY368150.1 | - | 15 - 16 | 1790 | 151 | |
BC013957.2 | - | 7 - 8 | 512 | 151 | |
DQ993201.1 | - | 17 - 18 | 1808 | 151 |