The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020728.2 | NP_065779.1 | 2 - 3 | 584 | 102 |
GenBank mRNA | AB033054.2 | - | 3 - 4 | 370 | 102 |
AK126214.1 | - | 2 - 3 | 229 | 102 | |
AK308824.1 | - | 2 - 3 | 192 | 102 | |
AL833233.1 | - | 2 - 3 | 193 | 102 | |
AY368150.1 | - | 2 - 3 | 584 | 102 | |
DQ993201.1 | - | 4 - 5 | 602 | 102 |