The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290184.1 | NP_001277113.1 | 11 - 12 | 1444 | 100 |
NM_001290186.1 | NP_001277115.1 | 7 - 8 | 891 | 100 | |
NM_015331.2 | NP_056146.1 | 10 - 11 | 1305 | 100 | |
XM_011509363.2 | XP_011507665.1 | 10 - 11 | 1522 | 100 | |
GenBank mRNA | AF240468.1 | - | 10 - 11 | 1323 | 100 |
AK296153.1 | - | 5 - 6 | 590 | 100 | |
AK314764.1 | - | 10 - 11 | 1212 | 100 | |
AY359120.1 | - | 10 - 11 | 1182 | 100 | |
BC047621.1 | - | 11 - 12 | 1401 | 100 | |
D87442.1 | - | 10 - 11 | 1179 | 100 |