Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCSTN
Entrez Gene ID
23385
Gene Name
nicastrin
Gene Aliases
ATAG1874
Chromosome Location
Chr.1: 160343056 - 160358952 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290184.1 NP_001277113.1 17 - 18 2273 81
NM_001290186.1 NP_001277115.1 13 - 14 1720 81
NM_015331.2 NP_056146.1 16 - 17 2134 81
GenBank mRNA AF240468.1 - 16 - 17 2152 81
AK296153.1 - 11 - 12 1419 81
AK299142.1 - 13 - 14 1607 81
AK314764.1 - 16 - 17 2041 81
AY359120.1 - 16 - 17 2011 81
BC047621.1 - 17 - 18 2230 81
D87442.1 - 16 - 17 2008 81

View Full Product Details