Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCSTN
Entrez Gene ID
23385
Gene Name
nicastrin
Gene Aliases
ATAG1874
Chromosome Location
Chr.1: 160343056 - 160358952 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290184.1 NP_001277113.1 6 - 7 850 115
NM_015331.2 NP_056146.1 5 - 6 711 115
XM_005245053.4 XP_005245110.1 5 - 6 928 115
XM_011509363.2 XP_011507665.1 5 - 6 928 115
GenBank mRNA AF240468.1 - 5 - 6 729 115
AK314764.1 - 5 - 6 618 115
AY359120.1 - 5 - 6 588 115
BC047621.1 - 6 - 7 807 115
D87442.1 - 5 - 6 585 115

View Full Product Details