The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290184.1 | NP_001277113.1 | 10 - 11 | 1362 | 104 |
NM_001290186.1 | NP_001277115.1 | 6 - 7 | 809 | 104 | |
NM_015331.2 | NP_056146.1 | 9 - 10 | 1223 | 104 | |
XM_005245053.4 | XP_005245110.1 | 9 - 10 | 1440 | 104 | |
XM_011509363.2 | XP_011507665.1 | 9 - 10 | 1440 | 104 | |
GenBank mRNA | AF240468.1 | - | 9 - 10 | 1241 | 104 |
AK296153.1 | - | 4 - 5 | 508 | 104 | |
AK314764.1 | - | 9 - 10 | 1130 | 104 | |
AY359120.1 | - | 9 - 10 | 1100 | 104 | |
BC047621.1 | - | 10 - 11 | 1319 | 104 | |
D87442.1 | - | 9 - 10 | 1097 | 104 |