The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257197.1 | NP_001244126.1 | 13 - 14 | 2224 | 88 |
NM_001257198.1 | NP_001244127.1 | 14 - 15 | 2105 | 88 | |
NM_003590.4 | NP_003581.1 | 14 - 15 | 2422 | 88 | |
XM_006712800.2 | XP_006712863.2 | 14 - 15 | 2095 | 88 | |
XM_011511994.2 | XP_011510296.1 | 13 - 14 | 1979 | 88 | |
XM_011511995.1 | XP_011510297.1 | 14 - 15 | 2007 | 88 | |
XM_011511996.2 | XP_011510298.1 | 14 - 15 | 2071 | 88 | |
GenBank mRNA | AB014517.2 | - | 14 - 15 | 2350 | 88 |
AF052147.1 | - | 7 - 8 | 845 | 88 | |
AF062537.1 | - | 14 - 15 | 2123 | 88 | |
AF064087.1 | - | 14 - 15 | 2115 | 88 | |
AK291151.1 | - | 14 - 15 | 2072 | 88 | |
AK299644.1 | - | 13 - 14 | 1834 | 88 | |
AK308496.1 | - | 14 - 15 | 2105 | 88 | |
BC031844.1 | - | 7 - 8 | 850 | 88 | |
BC039598.1 | - | 14 - 15 | 2367 | 88 | |
BC092409.1 | - | 14 - 15 | 2367 | 88 | |
U58089.1 | - | 10 - 11 | 1461 | 88 |