Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CUL3
Entrez Gene ID
8452
Gene Name
cullin 3
Gene Aliases
CUL-3, PHA2E
Chromosome Location
Chr.2: 224470150 - 224585397 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257197.1 NP_001244126.1 2 - 3 564 125
NM_001257198.1 NP_001244127.1 3 - 4 445 125
NM_003590.4 NP_003581.1 3 - 4 762 125
XM_006712800.2 XP_006712863.2 3 - 4 435 125
XM_011511995.1 XP_011510297.1 3 - 4 347 125
XM_011511996.2 XP_011510298.1 3 - 4 411 125
GenBank mRNA AB014517.2 - 3 - 4 690 125
AF062537.1 - 3 - 4 463 125
AF064087.1 - 3 - 4 455 125
AK291151.1 - 3 - 4 412 125
AK299644.1 - 2 - 3 172 125
AK308496.1 - 3 - 4 445 125
AY337761.1 - 3 - 4 908 125
BC039598.1 - 3 - 4 707 125
BC092409.1 - 3 - 4 707 125

View Full Product Details