The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021095.2 | NP_066918.2 | 15 - 16 | 2147 | 73 |
NR_028323.1 | 14 - 15 | 2499 | 73 | ||
XM_006712128.1 | XP_006712191.1 | 15 - 16 | 1942 | 73 | |
XM_006712129.1 | XP_006712192.1 | 15 - 16 | 1990 | 73 | |
XM_006712130.1 | XP_006712193.1 | 15 - 16 | 1981 | 73 | |
XM_011533146.2 | XP_011531448.1 | 13 - 14 | 1707 | 73 | |
XM_017005216.1 | XP_016860705.1 | 12 - 13 | 1717 | 73 | |
GenBank mRNA | AF069307.1 | - | 15 - 16 | 2043 | 73 |
AF081571.1 | - | 14 - 15 | 1859 | 73 | |
AK027855.1 | - | 14 - 15 | 2339 | 73 | |
AK075078.1 | - | 6 - 7 | 585 | 73 | |
AK293162.1 | - | 9 - 10 | 1031 | 73 | |
AK314545.1 | - | 16 - 17 | 1974 | 73 | |
BC012806.2 | - | 13 - 14 | 1709 | 73 | |
BC015631.2 | - | 14 - 15 | 1860 | 73 | |
BQ651664.1 | - | 3 - 4 | 386 | 73 |