The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033581.1 | NP_001028753.1 | 7 - 8 | 923 | 66 |
NM_001033582.1 | NP_001028754.1 | 7 - 8 | 820 | 66 | |
NM_001242874.1 | NP_001229803.1 | 7 - 8 | 930 | 66 | |
NM_002744.4 | NP_002735.3 | 10 - 11 | 1135 | 66 | |
XM_011541773.1 | XP_011540075.1 | 10 - 11 | 1131 | 66 | |
XM_011541774.1 | XP_011540076.1 | 7 - 8 | 996 | 66 | |
XM_011541776.1 | XP_011540078.1 | 7 - 8 | 936 | 66 | |
XM_011541778.1 | XP_011540080.1 | 7 - 8 | 849 | 66 | |
XM_017001791.1 | XP_016857280.1 | 7 - 8 | 780 | 66 | |
XM_017001799.1 | XP_016857288.1 | 7 - 8 | 1257 | 66 | |
XM_017001800.1 | XP_016857289.1 | 5 - 6 | 577 | 66 | |
XM_017001801.1 | XP_016857290.1 | 5 - 6 | 577 | 66 | |
XM_017001802.1 | XP_016857291.1 | 7 - 8 | 1133 | 66 | |
XM_017001804.1 | XP_016857293.1 | 7 - 8 | 1043 | 66 | |
XM_017001805.1 | XP_016857294.1 | 7 - 8 | 1013 | 66 | |
GenBank mRNA | AK097627.1 | - | 5 - 6 | 556 | 66 |
AK290995.1 | - | 10 - 11 | 974 | 66 | |
AK294649.1 | - | 7 - 8 | 925 | 66 | |
AK294782.1 | - | 7 - 8 | 843 | 66 | |
AK302160.1 | - | 5 - 6 | 547 | 66 | |
AL542576.3 | - | 7 - 8 | 810 | 66 | |
BC008058.2 | - | 10 - 11 | 1082 | 66 | |
BC014270.2 | - | 10 - 11 | 1135 | 66 | |
BT007082.1 | - | 10 - 11 | 974 | 66 | |
L14283.1 | - | 10 - 11 | 974 | 66 | |
Z15108.1 | - | 10 - 11 | 956 | 66 |