Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKCZ
Entrez Gene ID
5590
Gene Name
protein kinase C zeta
Gene Aliases
PKC-ZETA, PKC2
Chromosome Location
Chr.1: 2050470 - 2185395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033581.1 NP_001028753.1 10 - 11 1236 107
NM_001033582.1 NP_001028754.1 10 - 11 1133 107
NM_001242874.1 NP_001229803.1 10 - 11 1243 107
NM_002744.4 NP_002735.3 13 - 14 1448 107
XM_011541773.1 XP_011540075.1 13 - 14 1444 107
XM_011541774.1 XP_011540076.1 10 - 11 1309 107
XM_011541776.1 XP_011540078.1 10 - 11 1249 107
XM_011541778.1 XP_011540080.1 10 - 11 1162 107
XM_017001789.1 XP_016857278.1 13 - 14 1468 107
XM_017001790.1 XP_016857279.1 13 - 14 1468 107
XM_017001791.1 XP_016857280.1 10 - 11 1093 107
XM_017001792.1 XP_016857281.1 10 - 11 1273 107
XM_017001793.1 XP_016857282.1 10 - 11 1531 107
XM_017001794.1 XP_016857283.1 10 - 11 1186 107
XM_017001795.1 XP_016857284.1 10 - 11 1190 107
XM_017001796.1 XP_016857285.1 10 - 11 1273 107
XM_017001797.1 XP_016857286.1 8 - 9 914 107
XM_017001798.1 XP_016857287.1 10 - 11 1186 107
XM_017001799.1 XP_016857288.1 10 - 11 1570 107
XM_017001800.1 XP_016857289.1 8 - 9 890 107
XM_017001801.1 XP_016857290.1 8 - 9 890 107
XM_017001802.1 XP_016857291.1 10 - 11 1446 107
XM_017001804.1 XP_016857293.1 10 - 11 1356 107
XM_017001805.1 XP_016857294.1 10 - 11 1326 107
GenBank mRNA AK097627.1 - 8 - 9 869 107
AK131526.1 - 3 - 4 983 107
AK290995.1 - 13 - 14 1287 107
AK294649.1 - 10 - 11 1238 107
AK294782.1 - 10 - 11 1156 107
AK302160.1 - 8 - 9 860 107
BC008058.2 - 13 - 14 1395 107
BC014270.2 - 13 - 14 1448 107
BT007082.1 - 13 - 14 1287 107
L14283.1 - 13 - 14 1287 107
Z15108.1 - 13 - 14 1269 107

View Full Product Details