The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004664.3 | NP_004655.1 | 1 - 2 | 276 | 114 |
NR_136888.1 | 1 - 2 | 276 | 114 | ||
XM_011538928.2 | XP_011537230.1 | 1 - 2 | 270 | 114 | |
GenBank mRNA | AF087693.1 | - | 1 - 2 | 245 | 114 |
AF173081.1 | - | 1 - 2 | 276 | 114 | |
AK309256.1 | - | 1 - 2 | 65 | 114 | |
AK315321.1 | - | 1 - 2 | 120 | 114 | |
BC062332.1 | - | 1 - 2 | 141 | 114 | |
BC099921.1 | - | 1 - 2 | 86 | 114 | |
BC118609.1 | - | 1 - 2 | 220 | 114 | |
BC122561.1 | - | 1 - 2 | 220 | 114 | |
CR407680.1 | - | 1 - 2 | 83 | 114 |