Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D4
Entrez Gene ID
9882
Gene Name
TBC1 domain family member 4
Gene Aliases
AS160, NIDDM5
Chromosome Location
Chr.13: 75284664 - 75482646 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014832.4 NP_055647.2 11 - 12 2597 129
XM_005266603.1 XP_005266660.1 10 - 11 3000 129
XM_005266605.2 XP_005266662.1 11 - 12 1925 129
XM_006719903.3 XP_006719966.1 11 - 12 2285 129
XM_011535331.1 XP_011533633.1 10 - 11 2964 129
GenBank mRNA AB011175.1 - 11 - 12 2543 129
AB449885.1 - 11 - 12 2196 129
BC151239.1 - 11 - 12 2535 129

View Full Product Details