The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014832.4 | NP_055647.2 | 12 - 13 | 2625 | 154 |
XM_005266603.1 | XP_005266660.1 | 11 - 12 | 3028 | 154 | |
XM_005266605.2 | XP_005266662.1 | 12 - 13 | 1953 | 154 | |
XM_006719903.3 | XP_006719966.1 | 12 - 13 | 2313 | 154 | |
XM_011535331.1 | XP_011533633.1 | 11 - 12 | 2992 | 154 | |
GenBank mRNA | AB011175.1 | - | 12 - 13 | 2571 | 154 |
AB449885.1 | - | 12 - 13 | 2224 | 154 | |
BC151239.1 | - | 12 - 13 | 2563 | 154 |